Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054057.4 | NP_473398.1 | 6 | 613 | ||
GenBank mRNA | AK032308.1 | - | 6 | 563 | ||
AK134549.1 | - | 6 | 546 | |||
AK147670.1 | - | 5 | 2545 | |||
AK160172.1 | - | 6 | 567 | |||
AK182002.1 | - | 1 | 111 | |||
AK211128.1 | - | 2 | 236 | |||
AK218333.1 | - | 3 | 252 |