Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054078.2 | NP_473419.2 | 7 | 1402 | ||
GenBank mRNA | AJ309544.2 | - | 5 | 1136 | ||
AK133149.1 | - | 7 | 1402 | |||
AK140355.1 | - | 8 | 1926 | |||
AK155523.1 | - | 6 | 1420 | |||
BC020083.1 | - | 7 | 1476 | |||
BC048415.1 | - | 7 | 1405 |