Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054078.2 | NP_473419.2 | 9 | 1897 | ||
GenBank mRNA | AJ309544.2 | - | 7 | 1631 | ||
AK043800.1 | - | 1 | 113 | |||
AK133149.1 | - | 9 | 1897 | |||
AK140355.1 | - | 9 | 3397 | |||
AK155523.1 | - | 8 | 1915 | |||
BC020083.1 | - | 9 | 1971 | |||
BC038012.1 | - | 8 | 1913 | |||
BC048415.1 | - | 9 | 1900 |