Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142401.1 | NP_001135873.1 | 4, 5 | 577 | ||
NM_001142402.1 | NP_001135874.1 | 4, 5 | 559 | |||
NM_001142403.1 | NP_001135875.1 | 5, 6 | 616 | |||
NM_006016.4 | NP_006007.2 | 5, 6 | 616 | |||
GenBank mRNA | AF106518.1 | - | 4, 5 | 378 | ||
AF263279.1 | - | 5, 6 | 435 | |||
AF299341.1 | - | 5, 6 | 435 | |||
AF299342.1 | - | 4, 5 | 456 | |||
AF299343.1 | - | 4, 5 | 474 | |||
AK301692.1 | - | 4, 5 | 469 | |||
AK303525.1 | - | 4, 5 | 411 | |||
AK308664.1 | - | 4, 5 | 590 | |||
AK310854.1 | - | 5, 6 | 457 | |||
AK312357.1 | - | 5, 6 | 524 | |||
AK315908.1 | - | 4, 5 | 428 | |||
BC011522.2 | - | 5, 6 | 516 | 3854912 | ||
BC037914.1 | - | 4, 5 | 1118 | |||
BC040317.1 | - | 1, 2 | 2468 | |||
D14043.1 | - | 5, 6 | 514 | |||
FJ200494.1 | - | 5, 6 | 431 |