Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tirap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177845.1 | NP_001171316.1 | 6, 7 | 1204 | ||
NM_001177846.1 | NP_001171317.1 | 6, 7 | 1186 | |||
NM_001177847.1 | NP_001171318.1 | 6, 7 | 1132 | |||
NM_054096.2 | NP_473437.2 | 6, 7 | 1196 | |||
GenBank mRNA | AF378130.1 | - | 6, 7 | 1105 | ||
AF410784.1 | - | 5, 6 | 1028 | |||
AK036431.1 | - | 6, 7 | 1125 | |||
AK080135.1 | - | 6, 7 | 1205 | |||
AK151117.1 | - | 6, 7 | 1134 | |||
AK168494.1 | - | 6, 7 | 1124 | |||
BC005584.1 | - | 3, 4 | 862 | 3157913 |