Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip4k2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054097.3 | NP_473438.1 | 10 | 1997 | ||
GenBank mRNA | AB054591.1 | - | 10 | 1940 | ||
BC004670.1 | - | 1 | 297 | |||
BC005539.1 | - | 5 | 1211 | |||
BC021383.1 | - | 10 | 1930 | 5042933 |