Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054098.3 | NP_473439.2 | 2 | 513 | ||
GenBank mRNA | AJ319746.1 | - | 2 | 516 | ||
AK040760.1 | - | 2 | 529 | |||
AY029778.1 | - | 2 | 492 | |||
BC006651.1 | - | 2 | 513 | 3500056 |