Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078477.2 | NP_510962.2 | 2 | 2027 | ||
GenBank mRNA | AF283891.1 | - | 2 | 2026 | ||
AK154524.1 | - | 2 | 2054 | |||
AK167841.1 | - | 1 | 938 | |||
AK167940.1 | - | 1 | 938 | |||
BC021615.1 | - | 1 | 460 | |||
BC038918.1 | - | 1 | 472 |