Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166031.2 | NP_001159503.2 | 4 | 1447 | ||
NM_080452.3 | NP_536700.3 | 4 | 1504 | |||
GenBank mRNA | AK051184.1 | - | 4 | 1504 | ||
AK154131.1 | - | 4 | 1498 | |||
AK156412.1 | - | 3 | 1947 | |||
AK160129.1 | - | 4 | 1497 | |||
AK165956.1 | - | 4 | 1511 | |||
AK171699.1 | - | 3 | 1967 | |||
AK172120.1 | - | 3 | 1967 | |||
AK172432.1 | - | 3 | 1947 | |||
BC033460.1 | - | 3 | 1953 | |||
BC048377.1 | - | 4 | 1866 | |||
BC060503.1 | - | 4 | 1480 | 6306321 |