Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baalc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080640.5 | NP_542371.1 | 1 | 312 | ||
GenBank mRNA | AF371320.1 | - | 262 | |||
AK013358.1 | - | 282 | ||||
AK045383.1 | - | 1 | 327 | |||
AK079337.1 | - | 1 | 312 |