Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baalc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080640.5 | NP_542371.1 | 2 | 2406 | ||
GenBank mRNA | AK045383.1 | - | 2 | 2421 | ||
AK079337.1 | - | 2 | 2406 | |||
AK079550.1 | - | 1 | 2342 |