Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caskin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080643.2 | NP_542374.2 | 18 | 2319 | ||
GenBank mRNA | AF451978.1 | - | 17 | 2038 | ||
AK037133.1 | - | 5 | 444 | |||
AK129297.1 | - | 18 | 2357 | |||
BC053083.1 | - | 18 | 2319 | 5704642 |