Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006043.1 | NP_006034.1 | 2 | 1008 | ||
GenBank mRNA | AB209794.1 | - | 2 | 1061 | ||
AF105374.1 | - | 2 | 1008 | |||
AF105375.1 | - | 2 | 1008 | |||
AK313463.1 | - | 2 | 1344 | |||
AY359095.2 | - | 2 | 1371 | |||
BC093734.1 | - | 2 | 972 | 7939579 | ||
BC093736.1 | - | 2 | 971 | 7939581 |