Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170748.1 | NP_001164219.1 | 8 | 1049 | ||
NM_080856.4 | NP_543132.3 | 8 | 1049 | |||
GenBank mRNA | AF403042.1 | - | 3 | 466 | ||
AK052884.1 | - | 8 | 1051 | |||
AK164800.1 | - | 2 | 112 | |||
BC145002.1 | - | 3 | 435 |