Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_126166.4 | NP_569054.2 | 4 | 574 | ||
GenBank mRNA | AF355152.1 | - | 2 | 256 | ||
AF420279.1 | - | 1 | 213 | |||
AK083977.1 | - | 4 | 575 | |||
AK087739.1 | - | 3 | 396 | |||
AK166177.1 | - | 3 | 449 | |||
AK171438.1 | - | 4 | 557 | |||
BC099937.1 | - | 4 | 612 | 30084049 |