Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130796.4 | NP_570614.2 | 1 | 1204 | ||
GenBank mRNA | AF408408.1 | - | 1 | 1192 | ||
AK052343.1 | - | 1 | 1165 | |||
AK077890.1 | - | 1 | 798 | |||
AK143645.1 | - | 1 | 1199 | |||
AK155955.1 | - | 1 | 1197 | |||
BC063089.1 | - | 1 | 1204 | 6516923 | ||
BC137801.1 | - | 1 | 1190 | 8860820 | ||
BC145951.1 | - | 1 | 1190 | 40131254 |