Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130878.2 | NP_570948.1 | 17 | 4238 | ||
GenBank mRNA | AB093300.1 | - | 17 | 4654 | ||
AF426393.1 | - | 17 | 4125 | |||
AK143151.1 | - | 17 | 4179 | |||
BC042454.1 | - | 17 | 4148 | 5369576 | ||
BC049999.1 | - | 2 | 2202 | |||
BC058270.1 | - | 17 | 4238 | 6490597 |