Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168621.1 | NP_001162092.1 | 3, 4 | 1135 | ||
NM_001168622.1 | NP_001162093.1 | 2, 3 | 1039 | |||
NM_001168623.1 | NP_001162094.1 | 3, 4 | 334 | |||
NM_133206.3 | NP_573469.1 | 3, 4 | 1135 | |||
GenBank mRNA | AF378525.1 | - | 3, 4 | 840 | ||
AK030575.1 | - | 2, 3 | 185 | |||
AK076756.1 | - | 3, 4 | 282 | |||
AK078819.1 | - | 3, 4 | 361 | |||
AK079338.1 | - | 3, 4 | 1135 | |||
AK181114.1 | - | 1, 2 | 93 | |||
AK189177.1 | - | 1, 2 | 93 | |||
AK216619.1 | - | 3, 4 | 261 | |||
BC006765.1 | - | 3, 4 | 735 | 3710377 | ||
BC086770.1 | - | 3, 4 | 933 | 5720382 | ||
BM950431.1 | - | 2, 3 | 216 |