Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133207.2 | NP_573470.2 | 7 | 1398 | ||
GenBank mRNA | AJ291608.1 | - | 7 | 1205 | ||
AK034003.1 | - | 7 | 1399 | |||
AK052366.1 | - | 6 | 1261 |