Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133208.2 | NP_573471.2 | 1 | 726 | ||
GenBank mRNA | AF281141.1 | - | 1 | 726 | ||
AK029572.1 | - | 1 | 306 | |||
AK031950.1 | - | 1 | 2577 | |||
AK046006.1 | - | 2 | 758 | |||
AK133603.1 | - | 2 | 739 | |||
AK134983.1 | - | 1 | 304 | |||
BC117876.1 | - | 1 | 451 | 40095930 | ||
BC117877.1 | - | 1 | 451 | 40095933 |