Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133216.3 | NP_573479.3 | 8 | 824 | ||
GenBank mRNA | AF363970.1 | - | 8 | 764 | ||
AK146033.1 | - | 8 | 804 | |||
AK149676.1 | - | 8 | 763 | |||
AK158597.1 | - | 6 | 782 | |||
AK166715.1 | - | 8 | 724 | |||
BC065174.1 | - | 8 | 712 | 6415314 |