Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133216.3 | NP_573479.3 | 20 | 1919 | ||
GenBank mRNA | AF363970.1 | - | 20 | 1859 | ||
AK052094.1 | - | 12 | 1061 | |||
AK080423.1 | - | 9 | 667 | |||
AK146033.1 | - | 20 | 1899 | |||
AK149676.1 | - | 20 | 1858 | |||
AK158597.1 | - | 18 | 1876 | |||
AK166715.1 | - | 20 | 1819 | |||
AK169024.1 | - | 19 | 1745 | |||
AK177605.1 | - | 2 | 65 | |||
AK195402.1 | - | 2 | 79 | |||
AK197404.1 | - | 2 | 79 | |||
BC021534.1 | - | 11 | 854 | |||
BC065174.1 | - | 20 | 1807 | 6415314 |