Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133235.2 | NP_573498.1 | 2 | 297 | ||
GenBank mRNA | AK051807.1 | - | 2 | 328 | ||
AK051938.1 | - | 2 | 375 | |||
AK078160.1 | - | 2 | 377 | |||
BC066814.1 | - | 2 | 355 | |||
BC132117.1 | - | 2 | 287 | 40130394 | ||
BC132119.1 | - | 2 | 287 | 40130396 |