Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133235.2 | NP_573498.1 | 3 | 404 | ||
GenBank mRNA | AK051807.1 | - | 3 | 435 | ||
AK078160.1 | - | 3 | 484 | |||
BC066814.1 | - | 3 | 462 | |||
BC132117.1 | - | 3 | 394 | 40130394 | ||
BC132119.1 | - | 3 | 394 | 40130396 |