Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 19 | 2364 | ||
NM_001252486.1 | NP_001239415.1 | 19 | 2364 | |||
NM_133247.3 | NP_573510.2 | 19 | 2388 | |||
GenBank mRNA | AF383174.1 | - | 19 | 2377 | ||
AK036742.1 | - | 1 | 896 | |||
AK082251.1 | - | 19 | 2347 | |||
AK082943.1 | - | 19 | 2347 | |||
AK087467.1 | - | 19 | 2365 | |||
AK122442.1 | - | 16 | 4846 | |||
AK143844.1 | - | 19 | 2356 | |||
BC005506.1 | - | 19 | 2317 | |||
BC031366.1 | - | 11 | 1504 | |||
BC036980.1 | - | 12 | 1564 | |||
BC049870.1 | - | 6 | 629 | |||
BC067399.1 | - | 18 | 2230 | |||
BC089315.1 | - | 19 | 2310 |