Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286150.1 | NP_001273079.1 | 2 | 281 | ||
NM_006085.5 | NP_006076.4 | 2 | 281 | |||
XM_005272998.2 | XP_005273055.1 | 2 | 252 | |||
XM_005272999.4 | XP_005273056.1 | 2 | 361 | |||
XM_005273000.4 | XP_005273057.1 | 2 | 330 | |||
XM_005273004.2 | XP_005273061.1 | 2 | 252 | |||
XM_005273005.4 | XP_005273062.1 | 2 | 364 | |||
XM_006711114.3 | XP_006711177.1 | 2 | 333 | |||
XM_011509065.2 | XP_011507367.1 | 2 | 252 | |||
XM_017000043.1 | XP_016855532.1 | 2 | 353 | |||
XM_017000045.1 | XP_016855534.1 | 2 | 252 | |||
GenBank mRNA | AF125042.1 | - | 2 | 218 | ||
AJ249339.1 | - | 2 | 246 | |||
AK291943.1 | - | 2 | 216 | |||
AK300777.1 | - | 2 | 226 | |||
BC017801.1 | - | 2 | 220 | 4338207 | ||
BG428446.1 | - | 2 | 271 | |||
DC368603.1 | - | 2 | 281 | |||
EU794594.1 | - | 2 | 218 |