Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oosp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133353.3 | NP_579931.1 | 5 | 533 | ||
GenBank mRNA | AB050982.2 | - | 5 | 526 | ||
AB086437.1 | - | 4 | 418 | |||
AK033038.1 | - | 5 | 536 | |||
AK162132.1 | - | 5 | 536 | |||
BC100313.1 | - | 5 | 512 | 30936071 |