Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197181.1 | NP_001184110.1 | 3 | 445 | ||
NM_006086.3 | NP_006077.2 | 3 | 388 | |||
GenBank mRNA | AF427491.3 | - | 3 | 265 | ||
AK122757.1 | - | 2 | 2347 | |||
AK127418.1 | - | 443 | ||||
AK223039.1 | - | 3 | 322 | |||
AK292219.1 | - | 3 | 445 | |||
AK307201.1 | - | 4 | 461 | |||
AK314616.1 | - | 3 | 327 | |||
AU142509.1 | - | 3 | 325 | |||
BC000748.2 | - | 3 | 314 | 2823044 | ||
BC001678.2 | - | 2 | 133 | |||
BC003021.1 | - | 3 | 314 | 2823044 | ||
BC047518.1 | - | 1 | 78 | |||
CD621827.1 | - | 3 | 342 | |||
DA054466.1 | - | 3 | 388 | |||
S62643.1 | - | 3 | 244 | |||
U47634.1 | - | 3 | 265 |