Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159321.1 | NP_001152793.1 | 12 | 2304 | ||
NM_133355.1 | NP_579933.1 | 11 | 1608 | |||
GenBank mRNA | AF099933.2 | - | 11 | 1608 | ||
BC075624.1 | - | 1660 | 6841572 | |||
DQ193535.1 | - | 12 | 2304 |