Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133685.2 | NP_598446.2 | 6 | 662 | ||
GenBank mRNA | AB232617.1 | - | 6 | 454 | ||
AK007048.1 | - | 2 | 403 | |||
AK016608.1 | - | 2 | 392 | |||
AK051270.1 | - | 6 | 582 | |||
AK156677.1 | - | 6 | 663 | |||
AK159095.1 | - | 6 | 551 | |||
BC013063.1 | - | 6 | 591 | 3497938 |