Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133768.4 | NP_598529.1 | 8 | 665 | ||
GenBank mRNA | AK010922.1 | - | 8 | 678 | ||
AK076453.1 | - | 8 | 680 | |||
AK138423.1 | - | 8 | 662 | |||
AK140160.1 | - | 8 | 680 | |||
AK148764.1 | - | 8 | 649 | |||
AK168739.1 | - | 8 | 660 | |||
BC016670.1 | - | 8 | 665 | 2648753 |