Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133792.2 | NP_598553.1 | 5 | 699 | ||
GenBank mRNA | AF468958.1 | - | 5 | 611 | ||
AK085194.1 | - | 5 | 688 | |||
AK155004.1 | - | 5 | 695 | |||
AK163111.1 | - | 5 | 688 | |||
AK170814.1 | - | 5 | 687 | |||
BC019373.1 | - | 5 | 699 | 3964729 |