Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgdia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133796.7 | NP_598557.3 | 6 | 1412 | ||
GenBank mRNA | AK075656.1 | - | 5 | 1279 | ||
AK077343.1 | - | 6 | 1572 | |||
AK143516.1 | - | 6 | 1396 | |||
AK165241.1 | - | 6 | 1394 | |||
AK170553.1 | - | 6 | 1379 | |||
AK170718.1 | - | 6 | 1396 | |||
AK198117.1 | - | 1 | 210 | |||
AK206531.1 | - | 1 | 211 | |||
BC004732.1 | - | 6 | 1354 | 3583026 | ||
BC086755.1 | - | 6 | 1336 | 6414768 |