Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133819.3 | NP_598580.1 | 1 | 1222 | ||
GenBank mRNA | AK049028.1 | - | 1 | 926 | ||
AK082957.1 | - | 1 | 1058 | |||
AK086606.1 | - | 1 | 854 | |||
AK144678.1 | - | 1 | 975 | |||
AK146089.1 | - | 1 | 960 | |||
AK167565.1 | - | 1 | 1045 | |||
AK170862.1 | - | 1 | 1052 | |||
BC058078.1 | - | 1 | 1005 |