Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029842.1 | NP_001025013.1 | 6 | 2223 | ||
NM_134038.2 | NP_598799.1 | 6 | 2166 | |||
GenBank mRNA | AK032026.1 | - | 6 | 2226 | ||
AK170472.1 | - | 6 | 2089 | |||
BC017129.1 | - | 6 | 2157 | 4506255 |