Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142580.1 | NP_001136052.1 | 4 | 410 | ||
NM_001142581.1 | NP_001136053.1 | 4 | 410 | |||
NM_134044.3 | NP_598805.2 | 4 | 424 | |||
GenBank mRNA | AK038165.1 | - | 4 | 378 | ||
AK046322.1 | - | 4 | 381 | |||
AK049643.1 | - | 4 | 357 | |||
AK079055.1 | - | 4 | 357 | |||
AK082261.1 | - | 4 | 338 | |||
AK145043.1 | - | 393 | ||||
BB610654.1 | - | 4 | 411 | |||
BC016646.1 | - | 4 | 333 | 2651233 | ||
BC023716.1 | - | 4 | 435 | 5346489 |