Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253847.1 | NP_001240776.1 | 3 | 315 | ||
NM_001253848.1 | NP_001240777.1 | 3 | 303 | |||
NM_134048.2 | NP_598809.1 | 3 | 312 | |||
GenBank mRNA | AF167441.1 | - | 3 | 315 | ||
AK049141.1 | - | 3 | 251 | |||
AK051737.1 | - | 3 | 254 | |||
AK166017.1 | - | 3 | 245 | |||
BB651673.1 | - | 3 | 246 | |||
BC019529.1 | - | 3 | 236 | 4219836 |