Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134052.2 | NP_598813.1 | 4 | 930 | ||
GenBank mRNA | AK089966.1 | - | 4 | 914 | ||
AK145831.1 | - | 4 | 907 | |||
BC005695.1 | - | 4 | 929 | 3597652 |