Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134065.4 | NP_598826.3 | 3 | 1065 | ||
GenBank mRNA | AK034083.1 | - | 3 | 1034 | ||
AK038733.1 | - | 3 | 1040 | |||
AK046185.1 | - | 3 | 1040 | |||
AK160413.1 | - | 3 | 1000 | |||
AK194982.1 | - | 1 | 230 | |||
AK198088.1 | - | 1 | 230 | |||
BC014708.1 | - | 3 | 1020 | 4223134 |