Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005907.3 | NP_005898.2 | 12 | 2221 | ||
XM_005266986.4 | XP_005267043.1 | 12 | 2077 | |||
XM_011535833.2 | XP_011534135.1 | 11 | 1273 | |||
GenBank mRNA | AK025599.1 | - | 12 | 2216 |