Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134072.1 | NP_598833.1 | 3 | 309 | ||
GenBank mRNA | AK033537.1 | - | 3 | 327 | ||
AK136528.1 | - | 3 | 326 | |||
AY730283.1 | - | 3 | 271 | |||
BC013482.1 | - | 3 | 309 | 4240772 | ||
BC026628.1 | - | 3 | 308 | 4983237 |