Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134084.1 | NP_598845.1 | 5, 6 | 540 | ||
GenBank mRNA | AK028349.1 | - | 3, 4 | 2633 | ||
BC004041.1 | - | 5, 6 | 540 | 3496285 |