Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam19a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252310.1 | NP_001239239.1 | 4 | 903 | ||
NM_134096.2 | NP_598857.1 | 4 | 855 | |||
GenBank mRNA | AK090194.1 | - | 4 | 729 | ||
AK090307.1 | - | 4 | 905 | |||
AY325124.1 | - | 4 | 856 | |||
BC015306.1 | - | 4 | 856 | 4223181 | ||
BI986174.1 | - | 3 | 535 |