Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278595.1 | NP_001265524.1 | 3, 4 | 926 | ||
NM_005916.4 | NP_005907.3 | 3, 4 | 926 | |||
NM_182776.2 | NP_877577.1 | 3, 4 | 817 | |||
XM_005250348.3 | XP_005250405.1 | 3, 4 | 580 | |||
XM_017012217.1 | XP_016867706.1 | 3, 4 | 457 | |||
GenBank mRNA | AK055379.1 | - | 3, 4 | 794 | ||
AK096959.1 | - | 4, 5 | 290 | |||
AK226175.1 | - | 3, 4 | 356 | |||
AK293172.1 | - | 3, 4 | 328 | |||
AK314575.1 | - | 3, 4 | 393 | |||
BC009398.2 | - | 3, 4 | 344 | 4134871 | ||
BC013375.2 | - | 3, 4 | 362 | 3946342 | ||
D55716.1 | - | 3, 4 | 396 | |||
D86748.1 | - | 3, 4 | 366 | |||
KU178172.1 | - | 3, 4 | 280 | |||
KU178173.1 | - | 3, 4 | 280 |