Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134122.2 | NP_598883.1 | 4 | 1253 | ||
GenBank mRNA | BC021750.1 | - | 4 | 1256 | 4503599 | |
BC026576.1 | - | 4 | 1225 | 4984796 |