Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242389.1 | NP_001229318.1 | 7 | 662 | ||
NM_001242390.1 | NP_001229319.1 | 7 | 662 | |||
NM_001242391.1 | NP_001229320.1 | 7 | 662 | |||
NM_134125.4 | NP_598886.1 | 7 | 662 | |||
GenBank mRNA | AF502565.1 | - | 7 | 565 | ||
AK088909.1 | - | 7 | 612 | |||
AK149902.1 | - | 7 | 634 | |||
AY081142.1 | - | 7 | 613 | |||
BC003249.1 | - | 7 | 664 | 3500118 |