Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134130.1 | NP_598891.1 | 9 | 1363 | ||
GenBank mRNA | AF429302.1 | - | 9 | 1363 | ||
AK134676.1 | - | 9 | 1379 | |||
AK144238.1 | - | 9 | 1371 | |||
BC026770.1 | - | 9 | 1363 | 4234600 |