Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134135.1 | NP_598896.1 | 2 | 3341 | ||
GenBank mRNA | AK087519.1 | - | 4 | 3499 | ||
AK186098.1 | - | 1 | 129 | |||
AK190956.1 | - | 1 | 128 | |||
AK198355.1 | - | 1 | 128 | |||
AK216232.1 | - | 1 | 128 | |||
BC005502.1 | - | 2 | 3341 | 3490154 | ||
BC085175.1 | - | 4 | 3479 |