Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134139.1 | NP_598900.1 | 3, 4 | 307 | ||
GenBank mRNA | AK077521.1 | - | 3, 4 | 308 | ||
AK145704.1 | - | 3, 4 | 320 | |||
BC019968.1 | - | 3, 4 | 307 | 4167260 |