Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134151.4 | NP_598912.4 | 3 | 510 | ||
GenBank mRNA | AK028785.1 | - | 3 | 372 | ||
AK043837.1 | - | 3 | 434 | |||
AK076634.1 | - | 3 | 355 | |||
AK139579.1 | - | 3 | 355 | |||
AK145356.1 | - | 3 | 403 | |||
AK151880.1 | - | 3 | 362 | |||
AK169708.1 | - | 3 | 286 | |||
AK195910.1 | - | 2 | 144 | |||
AK198671.1 | - | 2 | 142 | |||
AK205652.1 | - | 2 | 143 | |||
BC013552.1 | - | 3 | 441 | 4236514 | ||
BC022143.1 | - | 3 | 405 | 5101324 | ||
BC026615.1 | - | 3 | 417 | 4976999 |